Stampa…

3-(2-Chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C2716-5G C2716-25G
TCIAC2716-5GEA 74 EUR
TCIAC2716-5G TCIAC2716-25G
3-(2-Chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one ≥98.0% (by GC, titration analysis)
3-(2-Chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
Formula: C₁₁H₁₁ClN₂O
Peso molecolare: 222,67 g/mol
MDL Number: MFCD03085747
CAS Number: 41078-70-0
EINECS: 255-207-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR