Stampa…

N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-(Benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine, 1-(N-Cbz-amidino)pyrazole, 1-[N-(Carbobenzoxy)amidino]pyrazole, N-Cbz-1H-pyrazole-1-carboxamidine, Benzyl (imino(1H-pyrazol-1-yl)methyl)carbamate

C2709-1G C2709-5G
TCIAC2709-1GEA 23 EUR
TCIAC2709-1G TCIAC2709-5G
N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine ≥98.0% (by HPLC, titration analysis)
N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine
Formula: C₁₂H₁₂N₄O₂
Peso molecolare: 244,25 g/mol
MDL Number: MFCD00274657
CAS Number: 152120-62-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR