Stampa…

3-Chloro-5-hydroxybenzonitrile ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C2708-1G C2708-5G
TCIAC2708-1GEA 72 EUR
TCIAC2708-1G TCIAC2708-5G
3-Chloro-5-hydroxybenzonitrile ≥95.0% (by GC)
3-Chloro-5-hydroxybenzonitrile
Formula: C₇H₄ClNO
Peso molecolare: 153,5658 g/mol
MDL Number: MFCD11226540
CAS Number: 473923-97-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR