Stampa…

4-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Chloro-3-(trifluoromethyl)benzene-1-sulphonyl chloride, 4-Chloro-3-(trifluoromethyl)benzenesulphonyl Chloride

C2705-1G C2705-5G
TCIAC2705-1GEA 79 EUR
TCIAC2705-1G TCIAC2705-5G
4-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
4-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃Cl₂F₃O₂S
Peso molecolare: 279,07 g/mol
Punto di ebollizione: 131°C
MDL Number: MFCD00024882
CAS Number: 32333-53-2
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Freezing point 26.0 to 30.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR