Stampa…

2-Chloro-4-fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Chloro-4-fluorobenzene-1-carbonyl chloride, 2-Chloro-4-fluorobenzoylchloride

C2697-5G C2697-25G
TCIAC2697-5GEA 32 EUR
TCIAC2697-5G TCIAC2697-25G
2-Chloro-4-fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
2-Chloro-4-fluorobenzoyl chloride
Formula: C₇H₃Cl₂FO
Peso molecolare: 193 g/mol
Punto di ebollizione: 95°C
Flash Pt: 107°C
MDL Number: MFCD00084954
CAS Number: 21900-54-9
UN: 3265
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR