1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
1-(Cbz-amino)cyclopropanecarboxylic acid, 1-(Benzyloxycarbonylamino)cyclopropanecarboxylic acid, 1-(Carbobenzoxyamino)cyclopropanecarboxylic acid, 1-(Z-Amino)cyclopropanecarboxylic acid
C2680-1G
C2680-5G
TCIAC2680-1GEA
33
EUR
TCIAC2680-1G
TCIAC2680-5G
1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid
Formula:
C₁₂H₁₃NO₄ Peso molecolare: 235,2359 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00083285 CAS Number: 84677-06-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...