Stampa…

2-Chloro-5-fluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

Synonyms: 2-Chloro-5-fluorobenzeneboronic acid

C2666-1G C2666-5G
TCIAC2666-1GEA 76 EUR
TCIAC2666-1G TCIAC2666-5G
2-Chloro-5-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-5-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MDL Number: MFCD06656272
CAS Number: 444666-39-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR