Chloromethyl chlorosulphate ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
CH₂Cl₂O₃S Punto di ebollizione: 50°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00041547 CAS Number: 49715-04-0 EINECS: 256-442-1 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...