Stampa…

3-Fluoropyridine-2-carbonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-Fluoro-2-pyridinecarbonitrile, 2-Cyano-3-fluoropyridine, 3-Fluoropicolinonitrile

C2655-1G C2655-5G
TCIAC2655-1GEA 23 EUR
TCIAC2655-1G TCIAC2655-5G
3-Fluoropyridine-2-carbonitrile ≥98.0% (by GC)
3-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
Peso molecolare: 122,1 g/mol
MDL Number: MFCD06797501
CAS Number: 97509-75-6
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR