2-Chloro-4-nitropyridine-N-oxide ≥96.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Chloro-4-nitropyridine N-oxide
C2654-5G
C2654-25G
TCIAC2654-5GEA
66
EUR
TCIAC2654-5G
TCIAC2654-25G
2-Chloro-4-nitropyridine-N-oxide ≥96.0% (by GC)
2-Chloro-4-nitropyridine-N-oxide
Formula:
C₅H₃ClN₂O₃ Peso molecolare: 174,54 g/mol |
MDL Number:
MFCD00955617 CAS Number: 14432-16-7 EINECS: 238-404-6 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...