Stampa…

6-Chloro-5-(2-chloroethyl)oxindole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloro-5-(2-chloroethyl)indolin-2-one, Ziprasidone Impurity F, 6-Chloro-5-(2-chloroethyl)-2-indolinone

C2651-5G C2651-1G
TCIAC2651-5GEA 148 EUR
TCIAC2651-5G TCIAC2651-1G
6-Chloro-5-(2-chloroethyl)oxindole ≥98.0% (by GC)
6-Chloro-5-(2-chloroethyl)oxindole
Formula: C₁₀H₉Cl₂NO
Peso molecolare: 230,09 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD03411598
CAS Number: 118289-55-7
EINECS: 421-320-0
UN: 3077
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 223.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR