Stampa…

ɑ-(4-Chlorophenyl)-2-pyridinemethanol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4-Chlorophenyl)(pyridin-2-yl)methanol, 2-[(4-Chlorophenyl)(hydroxy)methyl]pyridine

C2646-1G C2646-5G
TCIAC2646-1GEA 73 EUR
TCIAC2646-1G TCIAC2646-5G
ɑ-(4-Chlorophenyl)-2-pyridinemethanol ≥98.0% (by GC, titration analysis)
alpha-(4-Chlorophenyl)-2-pyridinemethanol
Formula: C₁₂H₁₀ClNO
Peso molecolare: 219,67 g/mol
MDL Number: MFCD00023493
CAS Number: 27652-89-7
EINECS: 248-592-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR