2-Chloro-4-methylpyrimidine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
C2631-5G
C2631-25G
TCIAC2631-5GEA
35
EUR
TCIAC2631-5G
TCIAC2631-25G
2-Chloro-4-methylpyrimidine ≥98.0% (by GC)
2-Chloro-4-methylpyrimidine
Formula:
C₅H₅ClN₂ Punto di ebollizione: 94°C |
MDL Number:
MFCD00054434 CAS Number: 13036-57-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...