Stampa…

Ceftizoxime ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

C2622-1G C2622-5G
TCIAC2622-1GEA 77 EUR
TCIAC2622-1G TCIAC2622-5G
Ceftizoxime ≥98.0% (by HPLC, titration analysis)
Ceftizoxime
Formula: C₁₃H₁₃N₅O₅S₂
Peso molecolare: 383,40 g/mol
Punto di fusione: 227°C
MDL Number: MFCD00072000
CAS Number: 68401-81-0
Merck Index: 14,01951

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +135.0 to +145.0 deg (C=1、0.1mol/LNaOH3mL+H2O7mL)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR