Stampa…

Clobetasol propionate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Cloβsol propionate, Clobetasol-17-propionate, 21-Chloro-9ɑ-fluoro-11β,17ɑ-dihydroxy-16β-methyl-1,4-pregnadiene-3,20-dione 17-Propionate

C2612-5G C2612-1G
TCIAC2612-5GEA 164 EUR
TCIAC2612-5G TCIAC2612-1G
Clobetasol propionate ≥98.0% (by HPLC, titration analysis)
Clobetasol propionate
Formula: C₂₅H₃₂ClFO₅
Peso molecolare: 466,97 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00058499
CAS Number: 25122-46-7
EINECS: 246-634-3
Merck Index: 14,02363

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 196.0 to 200.0 deg-C
Specific rotation [a]20/D +99.0 to +103.0 deg (C=1,Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR