Stampa…

Caronic anhydride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,3-Dimethyl-1,2-cyclopropanedicarboxylic Anhydride, 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione

C2596-1G C2596-5G
TCIAC2596-1GEA 76 EUR
TCIAC2596-1G TCIAC2596-5G
Caronic anhydride ≥98.0% (by GC, titration analysis)
Caronic anhydride
Formula: C₇H₈O₃
Peso molecolare: 140,14 g/mol
Punto di ebollizione: 137°C
MDL Number: MFCD09751198
CAS Number: 67911-21-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 98.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR