(R)-(-)-1-Cbz-3-Pyrrolidinol ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
C2585-5G
C2585-1G
TCIAC2585-5GEA
150
EUR
TCIAC2585-5G
TCIAC2585-1G
(R)-(-)-1-Cbz-3-Pyrrolidinol ≥98.0% (by GC)
(R)-(-)-1-Cbz-3-Pyrrolidinol
Formula:
C₁₂H₁₅NO₃ |
MDL Number:
MFCD07368258 CAS Number: 100858-33-1 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 79.0 deg-C |
Specific rotation [a]20/D | -21.0 to -25.0 deg (C=1, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...