3-Chlorobenzoylacetonitrile ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3'-Chloro-2-cyanoacetophenone, (3-Chlorobenzoyl)acetonitrile, 3-(3-Chlorophenyl)-3-oxopropanenitrile
C2581-1G
C2581-5G
TCIAC2581-1GEA
25
EUR
TCIAC2581-1G
TCIAC2581-5G
3-Chlorobenzoylacetonitrile ≥97.0% (by GC)
3-Chlorobenzoylacetonitrile
Formula:
C₉H₆ClNO Peso molecolare: 179,61 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00067891 CAS Number: 21667-62-9 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...