Corannulene ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Dibenzo[ghi,mno]fluoranthene
C2572-100MG
C2572-20MG
TCIAC2572-100MGEA
690
EUR
TCIAC2572-100MG
TCIAC2572-20MG
Corannulene ≥97.0% (by GC)
Corannulene
Formula:
C₂₀H₁₀ Peso molecolare: 250,30 g/mol Punto di fusione: 269°C |
MDL Number:
MFCD22124717 CAS Number: 5821-51-2 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...