2-Cloro-4-trifluorometilpiridina ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
C2568-1G
C2568-5G
TCIAC2568-1GEA
31
EUR
TCIAC2568-1G
TCIAC2568-5G
2-Cloro-4-trifluorometilpiridina ≥98.0% (by GC)
2-Cloro-4-trifluorometilpiridina
Formula:
C₆H₃ClF₃N Peso molecolare: 181,54 g/mol Punto di ebollizione: 147°C Flash Pt: 40°C |
MDL Number:
MFCD00042224 CAS Number: 81565-18-6 UN: 1992 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...