Stampa…

Cilnidipine ≥98.0% (by HPLC)

Fornitore: TCI

Synonyms: 3-Cinnamyl 5-(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-Methoxyethyl (2E)-3-Phenyl-2-propenyl Ester, 2-Methoxyethyl (2E)-3-Phenyl-2-propenyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

C2564-1G C2564-5G
TCIAC2564-1GEA 42 EUR
TCIAC2564-1G TCIAC2564-5G
Cilnidipine ≥98.0% (by HPLC)
Cilnidipine
Formula: C₂₇H₂₈N₂O₇
Peso molecolare: 492.53 g/mol
MDL Number: MFCD00865853
CAS Number: 132203-70-4
Merck Index: 14,02275

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.55 to 5.75 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR