Stampa…

2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chloromethyl-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride, Lansoprazole Chloromethyl Impurity

C2532-25G C2532-5G
TCIAC2532-25GEA 59 EUR
TCIAC2532-25G TCIAC2532-5G
2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
2-(Chloromethyl)-3-methyl-4-(2,2,2-trifluoroethoxy)pyridine hydrochloride
Formula: C₉H₉ClF₃NO
Peso molecolare: 276,08 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00800224
CAS Number: 127337-60-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR