Acido 3-cloro-2,4-difluorobenzoico ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₇H₃ClF₂O₂ |
MDL Number:
MFCD01631387 CAS Number: 154257-75-7 |
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 174.0 to 178.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...