Stampa…

Z-Hyp-OH ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: N-Benzyloxycarbonyl-4-hydroxy-L-proline, Z-L-4-Hydroxyproline, trans-N-Cbz-4-hydroxy-L-proline, (2S,4R)-1-((Benzyloxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, N-Cbz-Hydroxy-L-Proline

C2506-5G C2506-25G
TCIAC2506-5GEA 32 EUR
TCIAC2506-5G TCIAC2506-25G
Z-Hyp-OH ≥98.0% (by HPLC, titration analysis)
Z-Hyp-OH
Formula: C₁₃H₁₅NO₅
Peso molecolare: 265,3 g/mol
MDL Number: MFCD00037329
CAS Number: 13504-85-3
EINECS: 236-831-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C
Specific rotation [a]20/D -53.0 to -56.0 deg (C=2、EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR