Stampa…

5-Chloro-2-thiophenesulphonamide ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Chlorothiophene-2-sulphonamide, 2-Chloro-5-sulphamoylthiophene

C2441-1G C2441-5G
TCIAC2441-1GEA 42 EUR
TCIAC2441-1G TCIAC2441-5G
5-Chloro-2-thiophenesulphonamide ≥98.0% (by GC, titration analysis)
5-Chloro-2-thiophenesulphonamide
Formula: C₄H₄ClNO₂S₂
Peso molecolare: 197,65 g/mol
MDL Number: MFCD00052584
CAS Number: 53595-66-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR