Stampa…

Copper(II)-2,3,9,10,16,17,23,24-octafluorophthalocyanine ≥98.0% (by total nitrogen and titration analysis), purified by sublimation

Fornitore: TCI

C2427-1G C2427-100MG
TCIAC2427-1GEA 642 EUR
TCIAC2427-1G TCIAC2427-100MG
Copper(II)-2,3,9,10,16,17,23,24-octafluorophthalocyanine ≥98.0% (by total nitrogen and titration analysis), purified by sublimation
Copper(II)-2,3,9,10,16,17,23,24-octafluorophthalocyanine
Formula: C₃₂H₈CuF₈N₈
CAS Number: 148651-60-9

Visualizza i codici

Risultati del test di specificazione

Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR