Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Chloro[1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]copper(I), [1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]chlorocopper(I)
C2422-1G
C2422-200MG
TCIAC2422-1GEA
131
EUR
TCIAC2422-1G
TCIAC2422-200MG
Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) ≥97.0% (by titrimetric analysis)
Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I)
Formula:
C₂₁H₂₄ClCuN₂ Peso molecolare: 403,43 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD15144833 CAS Number: 873779-78-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...