Stampa…

Cyhalofop butyl ≥97.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Butyl (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionate, (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionic Acid Butyl Ester

C2417-1G C2417-25G
TCIAC2417-1GEA 33 EUR
TCIAC2417-1G TCIAC2417-25G
Cyhalofop butyl ≥97.0% (by GC)
Cyhalofop butyl
Formula: C₂₀H₂₀FNO₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01631151
CAS Number: 122008-85-9
UN: 3077
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR