ɑ-Cyclohexyl-DL-mandelic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₄H₁₈O₃ Punto di ebollizione: 135°C |
MDL Number:
MFCD00194543 CAS Number: 4335-77-7 EINECS: 224-380-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...