Stampa…

6-Chloro-2-benzoxazolinone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloro-1,3-benzoxazol-2(3H)-one, 6-Chlorobenzo[d]oxazol-2(3H)-one, 6-Chloro-2-hydroxybenzoxazole, 6-Chloro-2-benzoxazolone

C2378-25G C2378-5G
TCIAC2378-25GEA 165 EUR
TCIAC2378-25G TCIAC2378-5G
6-Chloro-2-benzoxazolinone ≥98.0% (by GC)
6-Chloro-2-benzoxazolinone
Formula: C₇H₄ClNO₂
Peso molecolare: 169,56 g/mol
MDL Number: MFCD00463914
CAS Number: 19932-84-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 193.0 to 197.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR