Stampa…

6-Chloro-3-hydroxypyridazine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-chloropyridazin-3-ol, 6-Chloro-3(2H)-pyridazinone

C2377-5G C2377-25G
TCIAC2377-5GEA 52 EUR
TCIAC2377-5G TCIAC2377-25G
6-Chloro-3-hydroxypyridazine ≥98.0% (by GC, titration analysis)
6-Chloro-3-hydroxypyridazine
Formula: C₄H₃ClN₂O
Peso molecolare: 130,53 g/mol
MDL Number: MFCD00233955
CAS Number: 19064-67-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 144.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR