Stampa…

Chromone-3-carboxylic acid ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C2367-1G C2367-5G
TCIAC2367-1GEA 26 EUR
TCIAC2367-1G TCIAC2367-5G
Chromone-3-carboxylic acid ≥98.0% (by GC)
Chromone-3-carboxylic acid
Formula: C₁₀H₆O₄
Peso molecolare: 190,15 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00017338
CAS Number: 39079-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b1267a96036130eb99e51406302a4a16f967822&vid=5b1267a96036130eb99e51406302a4a16f967822&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 198.0 to 203.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR