2-Chloro-4-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
C2349-1G
C2349-5G
TCIAC2349-1GEA
93
EUR
TCIAC2349-1G
TCIAC2349-5G
2-Chloro-4-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-4-hydroxybenzoic acid
Formula:
C₇H₅ClO₃ Peso molecolare: 172,57 g/mol Punto di fusione: 159°C |
MDL Number:
MFCD00045855 CAS Number: 56363-84-9 EINECS: 260-132-1 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...