7-Chloro-6-nitro-4-hydroxyquinazoline ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
7-Chloro-6-nitroquinazolin-4(3H)-one, 7-Chloro-6-nitro-4(3H)-quinazolinone, 7-Chloro-6-nitro-4-quinazolinol, 7-Chloro-6-nitro-4-quinazolinone
C2332-5G
C2332-25G
TCIAC2332-5GEA
59
EUR
TCIAC2332-5G
TCIAC2332-25G
7-Chloro-6-nitro-4-hydroxyquinazoline ≥98.0% (by titrimetric analysis)
7-Chloro-6-nitro-4-hydroxyquinazoline
Formula:
C₈H₄ClN₃O₃ Peso molecolare: 225,59 g/mol Punto di fusione: 303°C |
MDL Number:
MFCD09388771 CAS Number: 53449-14-2 |
Risultati del test di specificazione
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...