trans-1,4-Cyclohexanediol ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
trans-Cyclohexane-1,4-diol, trans-1,4-Dihydroxycyclohexane
C2320-1G
C2320-5G
TCIAC2320-1GEA
99
EUR
TCIAC2320-1G
TCIAC2320-5G
trans-1,4-Cyclohexanediol ≥97.0% (by GC)
trans-1,4-Cyclohexanediol
Formula:
C₆H₁₂O₂ Peso molecolare: 116,16 g/mol |
MDL Number:
MFCD00063612 CAS Number: 6995-79-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 138.0 to 143.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...