Stampa…

Copper(I)-2-thiophenecarboxylate

Fornitore: TCI EUROPE
Attenzione

Synonyms: (2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC

C2312-1G C2312-5G
TCIAC2312-1GEA 30 EUR
TCIAC2312-1G TCIAC2312-5G
Copper(I)-2-thiophenecarboxylate
Copper(I)-2-thiophenecarboxylate
Formula: C₅H₃CuO₂S
Peso molecolare: 190,68 g/mol
MDL Number: MFCD02183524
CAS Number: 68986-76-5
Merck Index: 14,02521

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystaline
Content (Copper) 30.0 to 40.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR