Stampa…

Chloroquine diphosphate salt ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate

C2301-25G C2301-500G C2301-100G
TCIAC2301-25GEA 43 EUR
TCIAC2301-25G TCIAC2301-500G TCIAC2301-100G
Chloroquine diphosphate salt ≥98.0% (by HPLC, titration analysis)
Chloroquine diphosphate salt
Formula: C₁₈H₂₆ClN₃
Peso molecolare: 515,86 g/mol
Punto di fusione: 199°C
MDL Number: MFCD00069852
CAS Number: 50-63-5
EINECS: 200-055-2
Merck Index: 14,02163

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR