Stampa…

(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetriol ≥98.0% (by GC)

Fornitore: TCI EUROPE

C2275-5G C2275-25G
TCIAC2275-5GEA 208 EUR
TCIAC2275-5G TCIAC2275-25G
(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetriol ≥98.0% (by GC)
(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetriol
Formula: C₆H₁₂O₃
Peso molecolare: 132,16 g/mol
MDL Number: MFCD00043062
CAS Number: 50409-12-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 185.0 to 189.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR