Chlorothiazide ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
6-Chloro-2H-1,2,4-benzothiadiazine-7-sulphonamide 1,1-Dioxide
C2259-25G
C2259-5G
TCIAC2259-25GEA
77
EUR
TCIAC2259-25G
TCIAC2259-5G
Chlorothiazide ≥97.0% (by HPLC, titration analysis)
Chlorothiazide
Formula:
C₇H₆ClN₃O₄S₂ Peso molecolare: 295,71 g/mol Punto di fusione: 348°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00058576 CAS Number: 58-94-6 EINECS: 200-404-9 Merck Index: 14,02168 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...