Stampa…

Cephalexin monohydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 7-(D-α-amino-phenylacetamido)-3-methyl-3-cepheme-4-carboxylic acid monohydrate, (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate

C2248-25G C2248-5G
TCIAC2248-25GEA 138 EUR
TCIAC2248-25G TCIAC2248-5G
Cephalexin monohydrate ≥98.0% (by HPLC, titration analysis)
Cephalexin monohydrate
Formula: C₁₆H₁₇N₃O₄S
Peso molecolare: 347,39 g/mol
MDL Number: MFCD00167148
CAS Number: 23325-78-2
Merck Index: 14,01974
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=343ebd38715d4da81c561f15df58709264103e68&vid=580fe915e532d4230cd5a9c8c2f7703ea6b2f1cc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +149 to +158 deg (C=0.5, H2O) (calcd.on anh.substance)
Water 4.0 to 8.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR