2-(2-Chlorophenyl)-4,5-diphenylimidazole ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
2-(2-Chlorophenyl)-4,5-diphenyl-1H-imidazole
C2240-25G
C2240-5G
TCIAC2240-25GEA
162
EUR
TCIAC2240-25G
TCIAC2240-5G
2-(2-Chlorophenyl)-4,5-diphenylimidazole ≥98.0% (by HPLC, titration analysis)
2-(2-Chlorophenyl)-4,5-diphenylimidazole
Formula:
C₂₁H₁₅ClN₂ Peso molecolare: 330,82 g/mol |
MDL Number:
MFCD00030328 CAS Number: 1707-67-1 EINECS: 216-951-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...