Stampa…

Cyclophosphamide monohydrate ≥98.0% (by total nitrogen basis)

Fornitore: TCI
Pericolo

Synonyms: 2-(Bis(2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide hydrate, Cyclophosphamide hydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-1,3,2-oxazaphosphorin-2-oxide

C2236-25G C2236-5G
TCIAC2236-25GEA 364 EUR
TCIAC2236-25G TCIAC2236-5G
Cyclophosphamide monohydrate ≥98.0% (by total nitrogen basis)
Cyclophosphamide monohydrate
Formula: C₇H₁₇Cl₂N₂O₃P
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00149395
CAS Number: 6055-19-2
EINECS: 200-015-4
UN: 3464
ADR: 6.1,III
Merck Index: 14,02747

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Melting point 49.0 to 53.0 deg-C
Water max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR