Folinic acid calcium salt, idrato ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₂₀H₂₁CaN₇O₇ |
MDL Number:
MFCD00006704 CAS Number: 1492-18-8 EINECS: 216-082-8 |
|
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Chelometric Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 17.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...