Stampa…

Cefotaxime sodium salt ≥96.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

C2224-25G C2224-5G
TCIAC2224-25GEA 251 EUR
TCIAC2224-25G TCIAC2224-5G
Cefotaxime sodium salt ≥96.0% (by HPLC)
Cefotaxime sodium salt
Formula: C₁₆H₁₇N₅O₇S₂
Peso molecolare: 477,44 g/mol
Punto di fusione: 163°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00079073
CAS Number: 64485-93-4
EINECS: 264-915-9
Merck Index: 14,01933

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D +58.0 to +64.0 deg (C=1, H2O) (calcd.on dried substance)
Drying loss max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR