Stampa…

(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 6-Chloropurine riboside

C2206-1G C2206-5G
TCIAC2206-1GEA 48 EUR
TCIAC2206-1G TCIAC2206-5G
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol ≥97.0% (by HPLC, titration analysis)
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
Formula: C₁₀H₁₁ClN₄O₄
Peso molecolare: 286,67 g/mol
MDL Number: MFCD00005738
CAS Number: 5399-87-1
EINECS: 226-438-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D -42.0 to -48.0 deg (C=0.8, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR