1,2,4,5-Cyclohexanetetracarboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Cyclohexane-1,2,4,5-tetracarboxylic acid
C2198-25G
C2198-5G
TCIAC2198-25GEA
383
EUR
TCIAC2198-25G
TCIAC2198-5G
1,2,4,5-Cyclohexanetetracarboxylic acid ≥98.0% (by GC, titration analysis)
1,2,4,5-Cyclohexanetetracarboxylic acid
Formula:
C₁₀H₁₂O₈ Peso molecolare: 260,20 g/mol Punto di fusione: 271°C |
MDL Number:
MFCD00435556 CAS Number: 15383-49-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...