Stampa…

3-Chloro-4-morpholino-1,2,5-thiadiazole ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Timolol Related Compound F, 4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine

C2189-25G C2189-5G
TCIAC2189-25GEA 131 EUR
TCIAC2189-25G TCIAC2189-5G
3-Chloro-4-morpholino-1,2,5-thiadiazole ≥98.0% (by GC)
3-Chloro-4-morpholino-1,2,5-thiadiazole
Formula: C₆H₈ClN₃OS
Peso molecolare: 205,66 g/mol
MDL Number: MFCD00274160
CAS Number: 30165-96-9
EINECS: 250-077-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR