Stampa…

Cesio carbonato ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

C2160-25G C2160-100G
TCIAC2160-25GEA 35 EUR
TCIAC2160-25G TCIAC2160-100G
Cesio carbonato ≥98.0% (by titrimetric analysis)
Cesio carbonato
Formula: Cs₂CO₃
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 14,02010

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR