Cbz-D-Ser-OH ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
(R)-2-(((Benzyloxy)carbonyl)amino)-3-hydroxypropanoic acid, N-Cbz-D-serine, Z-D-Ser-OH
C2137-1G
C2137-5G
TCIAC2137-1GEA
17
EUR
TCIAC2137-1G
TCIAC2137-5G
Cbz-D-Ser-OH ≥98.0% (by titrimetric analysis)
Cbz-D-Ser-OH
Formula:
C₁₁H₁₃NO₅ Peso molecolare: 239,2 g/mol |
MDL Number:
MFCD00063144 CAS Number: 6081-61-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 117.0 to 120.0 deg-C |
Specific rotation [a]20/D | -5.0 to -7.0 deg (C=6, AcOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...