Stampa…

5-Chloro-2,3-difluoropyridine ≥93.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C2113-25G C2113-5G
TCIAC2113-25GEA 100 EUR
TCIAC2113-25G TCIAC2113-5G
5-Chloro-2,3-difluoropyridine ≥93.0% (by GC)
5-Chloro-2,3-difluoropyridine
Formula: C5H2ClF2N
Peso molecolare: 149,5259 g/mol
MDL Number: MFCD04039314
CAS Number: 89402-43-7
EINECS: 410-090-7
UN: 1993
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR